6-Chloro-3-methoxypyridazin-4-amine 98%

Supplier: Ambeed
Warning

A812823-1G A812823-250MG A812823-25G
76880-338EA 54.11 USD
76880-338 76880-340 76880-342
6-Chloro-3-methoxypyridazin-4-amine 98%
6-Chloro-3-methoxypyridazin-4-amine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃O
MW: 159.574 g/mol
MDL Number: MFCD00234095
CAS Number: 14369-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR