6-tert-Butyl 3-ethyl 4,5-dihydro-1H-pyrazolo[3,4-c]pyridine-3,6(7H)-dicarboxylate 95%
Supplier: Ambeed
|
Warning
|
A812330-100MG
A812330-250MG
A812330-1G
76880-334EA
89.12
USD
76880-334
76880-336
77614-454
6-tert-Butyl 3-ethyl 4,5-dihydro-1H-pyrazolo[3,4-c]pyridine-3,6(7H)-dicarboxylate 95%
6-tert-Butyl 3-ethyl 4,5-dihydro-1H-pyrazolo[3,4-c]pyridine-3,6(7H)-dicarboxylate
Precaución: for Research Use Only
Formula:
C₁₄H₂₁N₃O₄ MW: 295.33 g/mol |
MDL Number:
MFCD12198787 CAS Number: 821785-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...