N-Phenyl-1-naphthylamine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Anilinonaphthalene
A812108-1000G
A812108-100G
A812108-25G
A812108-500G
76880-326EA
139
USD
76880-326
76880-328
76880-330
76880-332
N-Phenyl-1-naphthylamine 98%
N-Phenyl-1-naphthylamine
Precaución: for Research Use Only
Formula:
C₁₆H₁₃N |
MDL Number:
MFCD00003878 CAS Number: 90-30-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...