2,2,2-Trifluoro-1-(pyrazin-2-yl)ethanone 97%

Supplier: Ambeed
Warning

A812080-1G A812080-250MG
76880-322EA 427 USD
76880-322 76880-324
2,2,2-Trifluoro-1-(pyrazin-2-yl)ethanone 97%
2,2,2-Trifluoro-1-(pyrazin-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₃F₃N₂O
MDL Number: MFCD06657596
CAS Number: 1147103-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR