7-Bromo-2-indoleboronic acid 98%

Supplier: Ambeed
Warning

A811167-1G A811167-250MG A811167-5G
76880-316EA 188.99 USD
76880-316 76880-318 76880-320
7-Bromo-2-indoleboronic acid 98%
7-Bromo-2-indoleboronic acid

Precaución: for Research Use Only

Formula: C₈H₇BBrNO₂
MDL Number: MFCD09800883
CAS Number: 957120-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR