2-Bromo-4-hydroxynicotinic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-hydroxypyridine-3-carboxylic acid, 2-Bromo-3-carboxy-4-hydroxypyridine

A807987-1G A807987-250MG
76880-304EA 115.64 USD
76880-304 76880-306
2-Bromo-4-hydroxynicotinic acid 95%
2-Bromo-4-hydroxynicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄BrNO₃
MW: 218.005 g/mol
MDL Number: MFCD12026106
CAS Number: 1150561-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR