(3-Formyl-5-propoxyphenyl)boronic acid 95%

Supplier: Ambeed
Warning

A806107-100MG A806107-1G A806107-250MG
76880-288EA 16 USD
76880-288 76880-290 76880-292
(3-Formyl-5-propoxyphenyl)boronic acid 95%
(3-Formyl-5-propoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BO₄
MDL Number: MFCD07784397
CAS Number: 871125-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR