4-Cyano-1H-indole-2-boronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-4-carbonitrile

A801585-1G A801585-250MG A801585-5G
76880-280EA 217 USD
76880-280 76880-282 76880-284
4-Cyano-1H-indole-2-boronic acid, pinacol ester 98%
4-Cyano-1H-indole-2-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₁₇BN₂O₂
MW: 268.119 g/mol
MDL Number: MFCD11858395
CAS Number: 1256359-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR