1-Nitro-9,10-anthraquinone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Nitroanthraquinone, 1-Nitroanthracene-9,10-dione
A800523-100G
A800523-25G
A800523-500G
76880-266EA
23
USD
76880-266
76880-268
76880-270
1-Nitro-9,10-anthraquinone 98%
1-Nitro-9,10-anthraquinone
Precaución: for Research Use Only
Formula:
C₁₄H₇NO₄ MW: 253.2097 g/mol |
MDL Number:
MFCD00019140 CAS Number: 82-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...