3,5-Difluoro-4-methylaniline 98%

Supplier: Ambeed
Warning

A800291-1G A800291-250MG
76880-262EA 198.39 USD
76880-262 76880-264
3,5-Difluoro-4-methylaniline 98%
3,5-Difluoro-4-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇F₂N
MW: 143.13 g/mol
MDL Number: MFCD03094196
CAS Number: 878285-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR