CEP-18770 98%

Supplier: Ambeed
Warning

Synonyms: Delanzomib, ((R)-1-((2S,3R)-3-Hydroxy-2-(6-phenylpicolinamido)butanamido)-3-methylbutyl)boronic acid

A800043-100MG A800043-1MG A800043-5MG A800043-2MG A800043-50MG
76880-256EA 254.62 USD
76880-256 76880-258 76880-260 77674-162 77674-164
CEP-18770 98%
CEP-18770

Precaución: for Research Use Only

Formula: C₂₁H₂₈BN₃O₅
MW: 413.28 g/mol
MDL Number: MFCD18251439
CAS Number: 847499-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR