Ethyl 5-oxo-1-tosyl-2,3,4,5-tetrahydro-1H-benzo[b]azepine-4-carboxylate 95%

Supplier: Ambeed
Warning

A799481-100MG A799481-250MG
76880-252EA 65.78 USD
76880-252 76880-254
Ethyl 5-oxo-1-tosyl-2,3,4,5-tetrahydro-1H-benzo[b]azepine-4-carboxylate 95%
Ethyl 5-oxo-1-tosyl-2,3,4,5-tetrahydro-1H-benzo[b]azepine-4-carboxylate

Precaución: for Research Use Only

Formula: C₂₀H₂₁NO₅S
MW: 387.44 g/mol
MDL Number: MFCD11973934
CAS Number: 54620-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR