3-Propyl-1H-pyrazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A798195-100MG A798195-1G A798195-250MG A798195-5G
76880-242EA 15 USD
76880-242 76880-244 76880-246 76880-248
3-Propyl-1H-pyrazole-5-carboxylic acid 97%
3-Propyl-1H-pyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MDL Number: MFCD02169458
CAS Number: 76424-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR