2-(3-Bromo-5-propoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-propoxyphenylboronic acid, pinacol ester

A793953-1G A793953-250MG A793953-5G
76880-224EA 77.45 USD
76880-224 76880-226 76880-228
2-(3-Bromo-5-propoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(3-Bromo-5-propoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₅H₂₂BBrO₃
MW: 341.04 g/mol
MDL Number: MFCD12546585
CAS Number: 1218790-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR