6-Methylbenzo[d][1,3]dioxol-5-amine 95%

Supplier: Ambeed
Warning

A793459-100MG A793459-1G A793459-250MG A793459-5G
76880-216EA 118.83 USD
76880-216 76880-218 76880-220 76880-222
6-Methylbenzo[d][1,3]dioxol-5-amine 95%
6-Methylbenzo[d][1,3]dioxol-5-amine

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD00666599
CAS Number: 62052-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR