2-Methyl-5-(trifluoromethyl)benzaldehyde 98%

Supplier: Ambeed
Warning

A793369-1G A793369-250MG A793369-5G
76880-210EA 44 USD
76880-210 76880-212 76880-214
2-Methyl-5-(trifluoromethyl)benzaldehyde 98%
2-Methyl-5-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₇F₃O
MDL Number: MFCD06660200
CAS Number: 886498-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR