Benzo[d]thiazole-2-thiol 98+%

Supplier: Ambeed
Warning

A792447-5G A792447-25G
76880-208EA 7.43 USD
76880-208 77573-532
Benzo[d]thiazole-2-thiol 98+%
Benzo[d]thiazole-2-thiol

Precaución: for Research Use Only

Formula: C₇H₅NS₂
MDL Number: MFCD00005781
CAS Number: 118090-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR