3,5-Dichloro-2-nitrobenzoic acid 95%

Supplier: Ambeed
Danger

A787795-1G A787795-250MG A787795-5G
76880-200EA 45 USD
76880-200 76880-202 76880-204
3,5-Dichloro-2-nitrobenzoic acid 95%
3,5-Dichloro-2-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃Cl₂NO₄
MDL Number: MFCD01098205
CAS Number: 23082-45-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR