(4-((Trifluoromethyl)thio)phenyl)boronic acid 95+%

Supplier: Ambeed
Warning

A785945-100MG A785945-1G A785945-250MG A785945-50MG
76880-186EA 36.08 USD
76880-186 76880-188 76880-190 76880-192
(4-((Trifluoromethyl)thio)phenyl)boronic acid 95+%
(4-((Trifluoromethyl)thio)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₆BF₃O₂S
MW: 221.99 g/mol
MDL Number: MFCD12546499
CAS Number: 947533-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR