4-Chloro-6-methylpyridin-3-amine 95%

Supplier: Ambeed
Warning

A785430-100MG A785430-250MG
76880-176EA 169 USD
76880-176 76880-178
4-Chloro-6-methylpyridin-3-amine 95%
4-Chloro-6-methylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MW: 142.59 g/mol
MDL Number: MFCD11557208
CAS Number: 1092285-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR