(2R,3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-yl 2,2,2-trichloroacetimidate 98%

Supplier: Ambeed
Warning

A784629-100MG A784629-1G A784629-250MG
76880-174EA 35 USD
76880-174 77886-758 77886-759
(2R,3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-yl 2,2,2-trichloroacetimidate 98%
(2R,3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-yl 2,2,2-trichloroacetimidate

Precaución: for Research Use Only

Formula: C₃₆H₃₆Cl₃NO₆
MDL Number: MFCD03427010
CAS Number: 74808-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR