4-(Methylamino)piperidine dihydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Methylpiperidin-4-amine dihydrochloride
A784368-1G
A784368-250MG
A784368-5G
76880-168EA
19
USD
76880-168
76880-170
76880-172
4-(Methylamino)piperidine dihydrochloride 98%
4-(Methylamino)piperidine dihydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₄N₂·2ClH MW: 187.111 g/mol |
MDL Number:
MFCD04039797 CAS Number: 1220039-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...