6-Chloro-2-methylpyridazin-3(2H)-one 96%

Supplier: Ambeed
Warning

A781707-100MG A781707-250MG
76880-164EA 13.8 USD
76880-164 76880-166
6-Chloro-2-methylpyridazin-3(2H)-one 96%
6-Chloro-2-methylpyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O
MW: 144.55 g/mol
MDL Number: MFCD02178600
CAS Number: 10071-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR