4-[(2-Aminoethyl)carbamoyl]benzeneboronic acid, N2-BOC protected 98%

Supplier: Ambeed
Warning

Synonyms: (4-((2-((tert-Butoxycarbonyl)amino)ethyl)carbamoyl)phenyl)boronic acid, 4-[(2-BOC-Amino)ethylcarbamoyl]benzeneboronic acid

A780400-1G A780400-5G
76880-160EA 111.4 USD
76880-160 76880-162
4-[(2-Aminoethyl)carbamoyl]benzeneboronic acid, N2-BOC protected 98%
4-[(2-Aminoethyl)carbamoyl]benzeneboronic acid, N2-BOC protected

Precaución: for Research Use Only

Formula: C₁₄H₂₁BN₂O₅
MW: 308.138 g/mol
MDL Number: MFCD09258752
CAS Number: 860626-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR