(S)-3-Amino-2-pyrrolidinone Hydrochloride 98%

Supplier: Ambeed
Warning

A780238-100MG A780238-1G A780238-250MG
76880-154EA 26 USD
76880-154 76880-156 76880-158
(S)-3-Amino-2-pyrrolidinone Hydrochloride 98%
(S)-3-Amino-2-pyrrolidinone Hydrochloride

Precaución: for Research Use Only

Formula: C₄H₉ClN₂O
MDL Number: MFCD08273939
CAS Number: 56440-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR