4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzeneboronic acid, pinacol ester 95%

Supplier: Ambeed
Danger

Synonyms: 3,5-Dimethyl-1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1H-pyrazole, 3,5,-Dimethyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-pyrazole

A775829-1G A775829-250MG A775829-5G
76880-132EA 146 USD
76880-132 76880-134 76880-136
4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzeneboronic acid, pinacol ester 95%
4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₃BN₂O₂
MDL Number: MFCD09879979
CAS Number: 937796-06-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR