(3R,5S)-rel-3,5-Dimethylmorpholine hydrochloride 98%

Supplier: Ambeed
Warning

A775552-100MG A775552-250MG A775552-1G
76880-128EA 47.75 USD
76880-128 76880-130 77614-380
(3R,5S)-rel-3,5-Dimethylmorpholine hydrochloride 98%
(3R,5S)-rel-3,5-Dimethylmorpholine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO
MW: 151.63 g/mol
MDL Number: MFCD14525805
CAS Number: 154596-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR