[(Dimethylaminomethylidene)amino]-3H-1,2,4-dithiazoline-3-thione 95%

Supplier: Ambeed
Warning

A774142-1G A774142-25G A774142-5G
76880-126EA 10 USD
76880-126 77552-386 77552-388
[(Dimethylaminomethylidene)amino]-3H-1,2,4-dithiazoline-3-thione 95%
[(Dimethylaminomethylidene)amino]-3H-1,2,4-dithiazoline-3-thione

Precaución: for Research Use Only

Formula: C₅H₇N₃S₃
MDL Number: MFCD19704304
CAS Number: 1192027-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR