5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalic acid 96%

Supplier: Ambeed
Warning

A772031-1G A772031-5G A772031-25G
76880-114EA 27 USD
76880-114 76880-116 77884-711
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalic acid 96%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₇BO₆
MDL Number: MFCD11867793
CAS Number: 1041434-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR