6-Isopropylpyridin-2-amine 98%

Supplier: Ambeed
Warning

A769916-1G A769916-250MG A769916-5G
76880-096EA 47 USD
76880-096 76880-098 76880-100
6-Isopropylpyridin-2-amine 98%
6-Isopropylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂
MDL Number: MFCD09743891
CAS Number: 78177-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR