5-Bromothiazole-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A769531-100MG A769531-1G A769531-250MG A769531-5G
76880-092EA 83.82 USD
76880-092 76880-094 77552-372 77573-480
5-Bromothiazole-2-carboxylic acid 98%
5-Bromothiazole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₂BrNO₂S
MW: 208.03 g/mol
MDL Number: MFCD11521568
CAS Number: 957346-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR