N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Diethylamino)phenylboronic Acid Pinacol Ester, 2-[4-(Diethylamino)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A769524-1G A769524-250MG A769524-5G
76880-086EA 157.02 USD
76880-086 76880-088 76880-090
N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline 98%
N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

Precaución: for Research Use Only

Formula: C₁₆H₂₆BNO₂
MW: 275.20 g/mol
MDL Number: MFCD10000956
CAS Number: 920304-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR