4-Bromo-N-tert-butyl-3-(trifluoromethyl)benzenesulfonamide 96%

Supplier: Ambeed
Danger

A768027-100MG A768027-1G A768027-250MG
76880-078EA 7 USD
76880-078 76880-080 76880-082
4-Bromo-N-tert-butyl-3-(trifluoromethyl)benzenesulfonamide 96%
4-Bromo-N-tert-butyl-3-(trifluoromethyl)benzenesulphonamide

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrF₃NO₂S
MDL Number: MFCD09972162
CAS Number: 1020253-06-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR