2-Chloroethylammonium chloride 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloroethylamine hydrochloride, 2-Chloroethylamine HCl, 2-Chloroethanamine hydrochloride

A766373-100G A766373-10G A766373-25G A766373-500G A766373-5G A766373-1000G
76880-068EA 10.61 USD
76880-068 76880-070 76880-072 76880-074 76880-076 77595-216
2-Chloroethylammonium chloride 97%
2-Chloroethylammonium chloride

Precaución: for Research Use Only

Formula: C₂H₇Cl₂N
MW: 115.99 g/mol
MDL Number: MFCD00012887
CAS Number: 870-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR