(E)-2-Cyclopropylethylene-1-boronic acid, pinacol ester 98% stabilised

Supplier: Ambeed
Warning

Synonyms: (E)-2-Cyclopropylethene-1-boronic acid, (E)-2-(2-Cyclopropylvinyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, trans-2-Cyclopropylvinylboronic acid, pinacol ester

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A764966-25G A764966-5G A764966-1G
76880-064EA 293 USD
76880-064 76880-066 77595-212
(E)-2-Cyclopropylethylene-1-boronic acid, pinacol ester 98% stabilised
(E)-2-Cyclopropylethylene-1-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₉BO₂
MDL Number: MFCD07369776
CAS Number: 849061-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR