N-Biotinyl-N'-(3-maleimidopropionyl)-3,6-dioxaoctane-1,8-diamine 90%

Supplier: Ambeed
Warning

Synonyms: N-[2-[2-[2-[(3-Maleimidopropionyl)amino]ethoxy]ethoxy]ethyl]biotinamide, N-(2-(2-(2-(3-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanamido)ethoxy)ethoxy)ethyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide, Maleimide-PEG2-Biotin, Biotin-PEG2-Mal

A764213-100MG A764213-1G A764213-250MG
76880-056EA 57 USD
76880-056 77674-100 77674-102
N-Biotinyl-N'-(3-maleimidopropionyl)-3,6-dioxaoctane-1,8-diamine 90%
N-Biotinyl-N'-(3-maleimidopropionyl)-3,6-dioxaoctane-1,8-diamine

Precaución: for Research Use Only

Formula: C₂₃H₃₅N₅O₇S
MDL Number: MFCD09264743
CAS Number: 305372-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR