2,5-Dichloro-3-nitrobenzoic acid 95%

Supplier: Ambeed
Warning

A763637-1G A763637-250MG A763637-5G
76880-050EA 39.26 USD
76880-050 76880-052 76880-054
2,5-Dichloro-3-nitrobenzoic acid 95%
2,5-Dichloro-3-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃Cl₂NO₄
MW: 236.01 g/mol
MDL Number: MFCD00014691
CAS Number: 88-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR