4,4'-Bis(bromomethyl)-2,2'-bipyridine 98%

Supplier: Ambeed
Danger

A762678-100MG A762678-1G A762678-250MG A762678-5G
76880-036EA 28 USD
76880-036 76880-038 76880-040 76880-042
4,4'-Bis(bromomethyl)-2,2'-bipyridine 98%
4,4'-Bis(bromomethyl)-2,2'-bipyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀Br₂N₂
MW: 342.02 g/mol
MDL Number: MFCD16038220
CAS Number: 134457-14-0
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR