5-(Methoxycarbonyl)-1H-pyrrole-3-boronic acid, pinacol ester, N-BOC protected 98%

Supplier: Ambeed
Warning

Synonyms: 1-tert-Butyl 2-methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole-1,2-dicarboxylate

A761444-100MG A761444-1G A761444-250MG A761444-5G
76880-030EA 36.08 USD
76880-030 76880-032 76880-034 77573-460
5-(Methoxycarbonyl)-1H-pyrrole-3-boronic acid, pinacol ester, N-BOC protected 98%
5-(Methoxycarbonyl)-1H-pyrrole-3-boronic acid, pinacol ester, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₇H₂₆BNO₆
MW: 351.202 g/mol
MDL Number: MFCD11855974
CAS Number: 942070-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR