4-Chlororesorcinol 98%

Supplier: Ambeed
Warning

A761419-100G A761419-25G A761419-500G A761419-1000G
76880-024EA 15 USD
76880-024 76880-026 76880-028 77886-722
4-Chlororesorcinol 98%
4-Chlororesorcinol

Precaución: for Research Use Only

Formula: C₆H₅ClO₂
MDL Number: MFCD00002273
CAS Number: 95-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR