2-Chlorobenzamidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chlorobenzene-1-carboximidamide hydrochloride, 2-Chloro-benzamidine hydrochloride, 2-Chlorophenylamidine hydrochloride

A761079-1G A761079-5G A761079-250MG
76880-020EA 50 USD
76880-020 76880-022 77884-690
2-Chlorobenzamidine hydrochloride 97%
2-Chlorobenzamidine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈Cl₂N₂
MDL Number: MFCD00275607
CAS Number: 18637-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR