2-(Tetrahydro-2H-pyran-4-yl)acetamide 95%

Supplier: Ambeed
Warning

A758946-100MG A758946-1G A758946-250MG
76880-014EA 40.32 USD
76880-014 76880-016 76880-018
2-(Tetrahydro-2H-pyran-4-yl)acetamide 95%
2-(Tetrahydro-2H-pyran-4-yl)acetamide

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MW: 143.18 g/mol
MDL Number: MFCD11043432
CAS Number: 1314961-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR