Ethyl-2,4-dichloro-6-methylpyridine-3-carboxylate 98%
Supplier: Ambeed
|
Danger
|
A754793-10G
A754793-1G
A754793-25G
A754793-5G
A754793-100G
76880-004EA
55
USD
76880-004
76880-006
77674-070
77674-072
77884-679
Ethyl-2,4-dichloro-6-methylpyridine-3-carboxylate 98%
Ethyl-2,4-dichloro-6-methylpyridine-3-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₉Cl₂NO₂ |
MDL Number:
MFCD00173918 CAS Number: 86129-63-7 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...