4-Carboxy-3-fluorophenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

A751702-1G A751702-250MG A751702-5G A751702-25G
76879-992EA 25 USD
76879-992 76879-994 76879-996 77884-677
4-Carboxy-3-fluorophenylboronic acid pinacol ester 98%
4-Carboxy-3-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BFO₄
MDL Number: MFCD09027284
CAS Number: 867256-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR