(R)-3-Hydroxy-2-phenylpropanoic acid 95%

Supplier: Ambeed
Warning

A749272-100MG A749272-1G A749272-250MG
76879-982EA 97.61 USD
76879-982 76879-984 76879-986
(R)-3-Hydroxy-2-phenylpropanoic acid 95%
(R)-3-Hydroxy-2-phenylpropanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.17 g/mol
MDL Number: MFCD00211261
CAS Number: 17126-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR