(S)-tert-Butyl 6-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-aminohexanoate hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: H-Lys(Fmoc)-OtBu·HCl

A748404-1G A748404-25G A748404-5G
76879-968EA 16.98 USD
76879-968 76879-970 76879-972
(S)-tert-Butyl 6-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-aminohexanoate hydrochloride 95%
(S)-tert-Butyl 6-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-aminohexanoate hydrochloride

Precaución: for Research Use Only

Formula: C₂₅H₃₃ClN₂O₄
MW: 460.99 g/mol
MDL Number: MFCD26131502
CAS Number: 330795-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR