4-Bromo-3-chlorobenzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

A747731-10G A747731-1G A747731-5G
76879-962EA 50 USD
76879-962 76879-964 76879-966
4-Bromo-3-chlorobenzenesulfonyl chloride 98%
4-Bromo-3-chlorobenzenesulfonyl chloride

Precaución: for Research Use Only

Formula: C₆H₃BrCl₂O₂S
MDL Number: MFCD04973905
CAS Number: 874801-46-4
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR