(4-(3,5-Dimethyl-1H-pyrazol-1-yl)phenyl)boronic acid 96%

Supplier: Ambeed
Warning

A746111-1G A746111-250MG A746111-5G
76879-956EA 206 USD
76879-956 76879-958 76879-960
(4-(3,5-Dimethyl-1H-pyrazol-1-yl)phenyl)boronic acid 96%
(4-(3,5-Dimethyl-1H-pyrazol-1-yl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃BN₂O₂
MDL Number: MFCD08572133
CAS Number: 1025495-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR