4-(Diethylcarbamoyl)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-(N,N-Diethylaminocarbonyl)phenylboronic acid, pinacol ester, [4-(Diethylamine-1-carbonyl)phenyl]boronic acid pinacol ester, N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

A744520-1G A744520-25G A744520-5G
76879-950EA 15 USD
76879-950 76879-952 76879-954
4-(Diethylcarbamoyl)phenylboronic acid pinacol ester 98%
4-(Diethylcarbamoyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₆BNO₃
MDL Number: MFCD06795673
CAS Number: 325142-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR