5-Bromo-2-hydroxy-4-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-4-(trifluoromethyl)pyridin-2-ol

A740173-100MG A740173-1G A740173-250MG A740173-5G
76879-942EA 13.8 USD
76879-942 76879-944 76879-946 76879-948
5-Bromo-2-hydroxy-4-(trifluoromethyl)pyridine 98%
5-Bromo-2-hydroxy-4-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃BrF₃NO
MW: 241.9933 g/mol
MDL Number: MFCD11227189
CAS Number: 109919-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR